Gene Transcripts

Gene Symbol
Pik3ap1
Entrez Gene ID
83490
Gene Name
phosphoinositide-3-kinase adaptor protein 1
Gene Aliases
1810044J04Rik, BCAP
Chromosome Location
Chr. 19: 41272752 - 41385070 on Build GRCm38
UniGene ID
Mm.222266
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031376.3 NP_113553.1 6, 7 1087
GenBank mRNA AF293806.1 - 6, 7 1087
AK087722.1 - 4, 5 556
AK089785.1 - 6, 7 1120
AK150114.1 - 4, 5 556
AK153548.1 - 4, 5 538
AK154326.1 - 6, 7 1115
AK170130.1 - 6, 7 1120
AK170925.1 - 6, 7 1120
AK171021.1 - 6, 7 1115
AK172670.1 - 3, 4 1539
BC113141.1 - 6, 7 1046 40047744

View Full Product Details