Gene Transcripts

Gene Symbol
Pcdhga5
Entrez Gene ID
93713
Gene Name
protocadherin gamma subfamily A, 5
Gene Aliases
Chromosome Location
Chr. 18: 37694501 - 37841870 on Build GRCm38
UniGene ID
Mm.247203
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033588.4 NP_291066.1 1 742
GenBank mRNA AK043161.1 - 1 857
AK043491.1 - 1 819
AY013799.1 - 1 742
BC057404.1 - 1 842 6405582

View Full Product Details