Gene Transcripts

Gene Symbol
Glrx
Entrez Gene ID
93692
Gene Name
glutaredoxin
Gene Aliases
C86710, D13Wsu156e, Glrx1, Grx1, TTase
Chromosome Location
Chr. 13: 75839873 - 75850151 on Build GRCm38
UniGene ID
Mm.25844
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053108.4 NP_444338.2 1 252
GenBank mRNA AB013137.1 - 1 218
AF109314.1 - 1 228
AF276917.1 - 1 177
AK013722.1 - 1 234
AK153086.1 - 1 232
BC012642.1 - 1 229 4206114
BC054418.1 - 1 214
BY272278.1 - 1 271

View Full Product Details