Gene Transcripts

Gene Symbol
Emilin1
Entrez Gene ID
100952
Gene Name
elastin microfibril interfacer 1
Gene Aliases
5830419M17Rik, AW229038, BB105748, EMILIN-1, gp115
Chromosome Location
Chr. 5: 30913381 - 30921279 on Build GRCm38
UniGene ID
Mm.286375
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK088995.1 - 4 1262
AK155486.1 - 4 1500
BC005481.1 - 4 1198 3485057

View Full Product Details