Gene Transcripts

Gene Symbol
Obsl1
Entrez Gene ID
98733
Gene Name
obscurin-like 1
Gene Aliases
AW822216
Chromosome Location
Chr. 1: 75485825 - 75506452 on Build GRCm38
UniGene ID
Mm.236454
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178884.5 NP_849215.3 6 2512
GenBank mRNA AK029863.1 - 6 2301
AK048144.1 - 6 1704
AK162473.1 - 6 2422
BC051643.1 - 4 1064
BC056205.1 - 3 686
CA750706.1 - 2 431

View Full Product Details