Gene Transcripts

Gene Symbol
Psg21
Entrez Gene ID
72242
Gene Name
pregnancy-specific glycoprotein 21
Gene Aliases
1600019C01Rik, 1600025N01Rik, 1600026N13Rik, cea8
Chromosome Location
Chr. 7: 18646654 - 18656725 on Build GRCm38
UniGene ID
Mm.326858
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027403.4 NP_081679.2 5 1859
GenBank mRNA AA023284.1 - 1 27
AK005492.1 - 5 1857
AK005545.1 - 5 1502
BC100324.1 - 5 1836 30715195
BC100325.1 - 5 1836 30787148

View Full Product Details