Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028041.2 | NP_082317.1 | 14 | 1711 | |
GenBank mRNA | AF319547.1 | - | 14 | 2071 | |
AK028274.1 | - | 14 | 1725 | ||
AK031534.1 | - | 14 | 1721 | ||
AK031679.1 | - | 14 | 1729 | ||
AK147715.1 | - | 14 | 1726 | ||
AK152989.1 | - | 14 | 1727 | ||
AK155138.1 | - | 14 | 1723 | ||
AK167603.1 | - | 14 | 1725 | ||
AK168455.1 | - | 14 | 1725 | ||
AK169255.1 | - | 14 | 1724 | ||
AK170967.1 | - | 14 | 1723 | ||
AK184886.1 | - | 2 | 175 | ||
BC021900.1 | - | 5 | 716 | ||
BC043699.1 | - | 14 | 1711 | 5344158 | |
BC055026.1 | - | 10 | 1154 | ||
BC062920.1 | - | 14 | 1714 | ||
BC071251.1 | - | 8 | 1008 |