Gene Transcripts

Gene Symbol
Prcp
Entrez Gene ID
72461
Gene Name
prolylcarboxypeptidase (angiotensinase C)
Gene Aliases
2510048K03Rik, 2610104A14Rik, AI451719, HUMPCP, PCP
Chromosome Location
Chr. 7: 92875253 - 92934581 on Build GRCm38
UniGene ID
Mm.389969
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028243.3 NP_082519.1 3, 4 499
GenBank mRNA AK051677.1 - 3, 4 456
AK054462.1 - 3, 4 437
AK166282.1 - 3, 4 456
BC026424.1 - 3, 4 295
BC055022.1 - 3, 4 432 4017462
BC089575.1 - 3, 4 454

View Full Product Details