Gene Transcripts

Gene Symbol
Ankrd42
Entrez Gene ID
73845
Gene Name
ankyrin repeat domain 42
Gene Aliases
4931426M20, 4933417L02Rik, Ikbn
Chromosome Location
Chr. 7: 92584175 - 92637143 on Build GRCm38
UniGene ID
Mm.73248
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028665.4 NP_082941.2 4 1312
GenBank mRNA AK029888.1 - 4 1273
AK033599.1 - 4 1301
AK048925.1 - 4 1206
AK133339.1 - 1273
BC030885.1 - 4 941
BC100433.1 - 4 1306
BC145456.1 - 4 430 9054017
BC145457.1 - 4 430
BC145458.1 - 4 430

View Full Product Details