Gene Transcripts

Gene Symbol
Kbtbd12
Entrez Gene ID
74589
Gene Name
kelch repeat and BTB (POZ) domain containing 12
Gene Aliases
4833415F11Rik, 4933428M03Rik, Klhdc6
Chromosome Location
Chr. 6: 88547722 - 88627448 on Build GRCm38
UniGene ID
Mm.196226
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278671.1 NP_001265600.1 2 873
GenBank mRNA AK014702.1 - 2 870
AK016973.1 - 2 872
BC139158.1 - 1 818 8862180
BC139160.1 - 1 818 8862182

View Full Product Details