Gene Transcripts

Gene Symbol
Zfp689
Entrez Gene ID
71131
Gene Name
zinc finger protein 689
Gene Aliases
4933416E05Rik, Znf689
Chromosome Location
Chr. 7: 127442136 - 127449158 on Build GRCm38
UniGene ID
Mm.474533
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175163.3 NP_780372.2 3 1085
GenBank mRNA AK028417.1 - 3 1048
AK051667.1 - 3 899
BC052084.1 - 3 1085 6404620

View Full Product Details