Gene Transcripts
- Gene Symbol
- 4932414N04Rik
- Entrez Gene ID
- 75721
- Gene Name
- RIKEN cDNA 4932414N04 gene
- Gene Aliases
- Chromosome Location
- Chr. 2: 68656791 - 68748526 on Build GRCm38
- UniGene ID
- Mm.65456
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_183113.3 |
NP_898936.1
|
16 |
1362 |
|
GenBank mRNA |
AK016523.1 |
- |
16 |
1363 |
|
View Full Product Details