Gene Transcripts

Gene Symbol
Slc25a11
Entrez Gene ID
67863
Gene Name
solute carrier family 25 (mitochondrial carrier oxoglutarate carrier), member 11
Gene Aliases
2310022P18Rik, 2oxoc
Chromosome Location
Chr. 11: 70644019 - 70647423 on Build GRCm38
UniGene ID
Mm.296082
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024211.3 NP_077173.1 5 779
GenBank mRNA AK003957.1 - 4 418
AK009487.1 - 5 725
AK009824.1 - 5 679
AK168951.1 - 5 704
BC003455.1 - 5 755 2648336
BC019631.1 - 5 733 4023179

View Full Product Details