Gene Transcripts

Gene Symbol
Snx5
Entrez Gene ID
69178
Gene Name
sorting nexin 5
Gene Aliases
0910001N05Rik, 1810032P22Rik, AU019504, D2Ertd52e
Chromosome Location
Chr. 2: 144250123 - 144270902 on Build GRCm38
UniGene ID
Mm.273379
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199188.1 NP_001186117.1 4 490
NM_024225.5 NP_077187.1 3 401
GenBank mRNA AK007676.1 - 3 349
AK029463.1 - 3 361
AK031873.1 - 4 382
AK049129.1 - 4 382
AK078776.1 - 4 383
AK144710.1 - 4 383
AK152311.1 - 4 426
AK159904.1 - 3 306
AK167362.1 - 3 321
AK169545.1 - 3 622
AK169786.1 - 4 382
AK184438.1 - 2 214
AK187575.1 - 2 213
AK214265.1 - 2 214
BC002242.1 - 4 459 3492262

View Full Product Details