Gene Transcripts

Gene Symbol
Snx5
Entrez Gene ID
69178
Gene Name
sorting nexin 5
Gene Aliases
0910001N05Rik, 1810032P22Rik, AU019504, D2Ertd52e
Chromosome Location
Chr. 2: 144250123 - 144270902 on Build GRCm38
UniGene ID
Mm.273379
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199188.1 NP_001186117.1 12 1185
NM_024225.5 NP_077187.1 11 1096
GenBank mRNA AK007676.1 - 11 1044
AK029463.1 - 11 1056
AK031873.1 - 12 1077
AK049129.1 - 12 1077
AK078776.1 - 12 1078
AK144710.1 - 12 1081
AK152311.1 - 12 1122
AK159115.1 - 8 639
AK159904.1 - 11 1001
AK167362.1 - 11 1016
AK169545.1 - 10 1991
AK169786.1 - 12 1077
BC002242.1 - 12 1154 3492262

View Full Product Details