Gene Transcripts

Gene Symbol
Rmnd5a
Entrez Gene ID
68477
Gene Name
required for meiotic nuclear division 5 homolog A (S. cerevisiae)
Gene Aliases
1110007A06Rik, AJ237586, AW125795, Gid2, MTA.D02.090
Chromosome Location
Chr. 6: 71388634 - 71440637 on Build GRCm38
UniGene ID
Mm.28474
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024288.2 NP_077250.2 5 1143
GenBank mRNA AK003512.2 - 1 75
AK148556.1 - 4 378
AK149170.1 - 4 378
AK164384.1 - 5 1144
BC034085.1 - 1 129
BC050876.1 - 5 994 6512227

View Full Product Details