Gene Transcripts

Gene Symbol
Plxdc2
Entrez Gene ID
67448
Gene Name
plexin domain containing 2
Gene Aliases
1200007L24Rik, 5430431D22Rik, AI646728, AU022916, AV231634, Tem7r
Chromosome Location
Chr. 2: 16356210 - 16755839 on Build GRCm38
UniGene ID
Mm.313938
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026162.6 NP_080438.2 2 1083
GenBank mRNA AF378761.1 - 2 282
AK004640.1 - 2 1065
AK019482.2 - 2 1039
AK035165.1 - 2 1084
AK154921.1 - 2 1030
BC057881.1 - 2 343 5349869

View Full Product Details