Gene Transcripts
- Gene Symbol
- Ero1lb
- Entrez Gene ID
- 67475
- Gene Name
- ERO1-like beta (S. cerevisiae)
- Gene Aliases
- 1300013B24Rik, 1700065B09Rik, AI447560, ero1-beta
- Chromosome Location
- Chr. 13: 12565809 - 12609530 on Build GRCm38
- UniGene ID
- Mm.358706
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026184.2 |
NP_080460.2
|
5, 6 |
624 |
|
GenBank mRNA |
AF489856.1 |
- |
5, 6 |
430 |
|
AK004981.1 |
- |
|
601 |
|
AK006886.1 |
- |
5, 6 |
487 |
|
AK041951.1 |
- |
5, 6 |
600 |
|
AK160664.1 |
- |
5, 6 |
640 |
|
BC058721.1 |
- |
5, 6 |
624 |
5705507 |
BC111900.1 |
- |
5, 6 |
594 |
40062137 |
BC112426.1 |
- |
5, 6 |
594 |
40062136 |
BC145888.1 |
- |
5, 6 |
575 |
40131191 |
BC145890.1 |
- |
5, 6 |
575 |
40131193 |
View Full Product Details