Gene Transcripts

Gene Symbol
FSBP
Entrez Gene ID
100861412
Gene Name
fibrinogen silencer binding protein
Gene Aliases
Chromosome Location
Chr. 8: 94427712 - 94436952 on Build GRCh38
UniGene ID
Hs.30561
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256141.1 NP_001243070.1 1 313
GenBank mRNA AF007866.1 - 1 265

Gene Symbol
RAD54B
Entrez Gene ID
25788
Gene Name
RAD54 homolog B (S. cerevisiae)
Gene Aliases
RDH54
Chromosome Location
Chr. 8: 94371960 - 94475115 on Build GRCh38
UniGene ID
Hs.30561
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205263.1 NP_001192192.1 1 313
GenBank mRNA DA553267.1 - 1 313

View Full Product Details