Gene Transcripts

Gene Symbol
Ppid
Entrez Gene ID
67738
Gene Name
peptidylprolyl isomerase D (cyclophilin D)
Gene Aliases
4930564J03Rik, CYP-40, Ppidl, Ppif
Chromosome Location
Chr. 3: 79591389 - 79603650 on Build GRCm38
UniGene ID
Mm.295252
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026352.3 NP_080628.1 10 1674
GenBank mRNA AK003402.1 - 10 1631
AK016221.1 - 10 1669
AK150520.1 - 10 1689
AK151091.1 - 10 1689
BC019778.1 - 10 1674 4035785

View Full Product Details