Gene Transcripts

Gene Symbol
Mgarp
Entrez Gene ID
67749
Gene Name
mitochondria localized glutamic acid rich protein
Gene Aliases
4930583H14Rik, AI195347, CESP-1, HUMMR, Osap, Qsap
Chromosome Location
Chr. 3: 51388412 - 51396547 on Build GRCm38
UniGene ID
Mm.273339
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026358.3 NP_080634.2 4 506
GenBank mRNA AF228502.1 - 4 485
AK006339.1 - 4 468
AK016342.1 - 4 479
AK151401.1 - 4 500
BC012511.1 - 4 499 4500810
BC022740.1 - 4 660 4511440
BC049556.1 - 4 468 6772062

View Full Product Details