Gene Transcripts

Gene Symbol
Eif1b
Entrez Gene ID
68969
Gene Name
eukaryotic translation initiation factor 1B
Gene Aliases
1500010M16Rik, 3110001N14Rik, Gc20
Chromosome Location
Chr. 9: 120492606 - 120495327 on Build GRCm38
UniGene ID
Mm.28753
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026892.3 NP_081168.1 4 546
GenBank mRNA AK005192.1 - 4 513
AK013965.1 - 4 519
BC030319.1 - 4 415 5372299
BC033505.1 - 4 517 4209443

View Full Product Details