Gene Transcripts
- Gene Symbol
- Ebna1bp2
- Entrez Gene ID
- 69072
- Gene Name
- EBNA1 binding protein 2
- Gene Aliases
- 1810014B19Rik, AA537058, B830003A16Rik, C81321, Ebp2, Nobp, p40
- Chromosome Location
- Chr. 4: 118620799 - 118627776 on Build GRCm38
- UniGene ID
- Mm.29906
- Species
- Mus musculus
- Gene Symbol
- D4Ertd617e
- Entrez Gene ID
- 100041290
- Gene Name
- DNA segment, Chr 4, ERATO Doi 617, expressed
- Gene Aliases
- Chromosome Location
- Chr. 4: 118626404 - 118631915 on Build GRCm38
- UniGene ID
- Mm.472885
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK021029.1 |
- |
1 |
483 |
|
View Full Product Details