Gene Transcripts

Gene Symbol
Comtd1
Entrez Gene ID
69156
Gene Name
catechol-O-methyltransferase domain containing 1
Gene Aliases
1810030M08Rik, MT773
Chromosome Location
Chr. 14: 21845861 - 21848910 on Build GRCm38
UniGene ID
Mm.11827
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026965.2 NP_081241.1 7 1002
GenBank mRNA AK054334.1 - 7 1003

View Full Product Details