Gene Transcripts

Gene Symbol
Reep6
Entrez Gene ID
70335
Gene Name
receptor accessory protein 6
Gene Aliases
0610011M24Rik, Dp1l1
Chromosome Location
Chr. 10: 80330003 - 80336441 on Build GRCm38
UniGene ID
Mm.28147
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204931.1 NP_001191860.1 5 839
NM_139292.2 NP_647453.1 6 920
GenBank mRNA AB039933.1 - 6 895
BC029741.1 - 5 826 4195889

View Full Product Details