Gene Transcripts

Gene Symbol
Psmd11
Entrez Gene ID
69077
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 11
Gene Aliases
1700089D09Rik, 1810019E17Rik, 2610024G20Rik, 2810055C24Rik, C78232, P44.5, S9
Chromosome Location
Chr. 11: 80428615 - 80473248 on Build GRCm38
UniGene ID
Mm.260539
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178616.3 NP_848731.2 3 305
GenBank mRNA AK007547.1 - 3 274
AK012951.1 - 3 314
AK051738.1 - 3 295
AK076002.1 - 3 293
AK089942.1 - 3 270
AK136049.1 - 3 268
AK140773.1 - 289
AK167559.1 - 3 276
AK168289.1 - 3 297
AK214581.1 - 2 159
AK215818.1 - 2 159
BC037590.1 - 3 254
BC055457.1 - 3 237
BC090664.1 - 3 257 30677672
BC090980.1 - 3 272 30283128
BC100595.1 - 3 267 6767671
BC119136.1 - 3 276 8733885
BC119138.1 - 3 276 8733887
BY092952.1 - 3 306

View Full Product Details