Gene Transcripts

Gene Symbol
Cmbl
Entrez Gene ID
69574
Gene Name
carboxymethylenebutenolidase-like (Pseudomonas)
Gene Aliases
2310016A09Rik
Chromosome Location
Chr. 15: 31565374 - 31590119 on Build GRCm38
UniGene ID
Mm.28108
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181588.3 NP_853619.1 6 844
GenBank mRNA AK009374.1 - 5 836
BC024580.1 - 6 868 5064316

View Full Product Details