Gene Transcripts
- Gene Symbol
- Srrm3
- Entrez Gene ID
- 58212
- Gene Name
- serine/arginine repetitive matrix 3
- Gene Aliases
- 2900083I11Rik, AB041661, SRm300-like, Srrm2l
- Chromosome Location
- Chr. 5: 135806861 - 135874773 on Build GRCm38
- UniGene ID
- Mm.271230
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB041661.1 |
- |
3 |
594 |
|
View Full Product Details