Gene Transcripts
- Gene Symbol
- Nisch
- Entrez Gene ID
- 64652
- Gene Name
- nischarin
- Gene Aliases
- 1200007D05Rik, 3202002H23Rik, AW494485, I-1
- Chromosome Location
- Chr. 14: 31170928 - 31206826 on Build GRCm38
- UniGene ID
- Mm.298728
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022656.2 |
NP_073147.2
|
9, 10 |
1165 |
|
GenBank mRNA |
AF315344.1 |
- |
5, 6 |
600 |
|
AK004628.1 |
- |
9, 10 |
1002 |
|
AK036043.1 |
- |
9, 10 |
1020 |
|
AK049791.1 |
- |
9, 10 |
1020 |
|
AK122412.1 |
- |
9, 10 |
1140 |
|
AK160990.1 |
- |
9, 10 |
1039 |
|
BC003270.1 |
- |
9, 10 |
1027 |
|
BC066838.1 |
- |
9, 10 |
1023 |
|
BC089329.1 |
- |
4, 5 |
384 |
|
BC108364.1 |
- |
4, 5 |
360 |
|
BC144980.1 |
- |
5, 6 |
494 |
|
View Full Product Details