Gene Transcripts

Gene Symbol
Nisch
Entrez Gene ID
64652
Gene Name
nischarin
Gene Aliases
1200007D05Rik, 3202002H23Rik, AW494485, I-1
Chromosome Location
Chr. 14: 31170928 - 31206826 on Build GRCm38
UniGene ID
Mm.298728
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022656.2 NP_073147.2 13, 14 1703
GenBank mRNA AF315344.1 - 9, 10 1138
AK122412.1 - 11, 12 1219
BC089329.1 - 8, 9 922
BC108364.1 - 8, 9 898
BC144980.1 - 9, 10 1032

View Full Product Details