Gene Transcripts

Gene Symbol
Rnf128
Entrez Gene ID
66889
Gene Name
ring finger protein 128
Gene Aliases
1300002C13Rik, AI987883, GRAIL, Greul1
Chromosome Location
Chr. X: 139563340 - 139673145 on Build GRCm38
UniGene ID
Mm.27764
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023270.5 NP_075759.3 1 485
GenBank mRNA AB041548.1 - 1 485
AF426411.1 - 1 299
AK004847.1 - 1 486
AK008312.1 - 1 429
AK018582.1 - 1 487
AK146266.1 - 1 482
AK167031.1 - 1 471
AY112656.1 - 1 472
BC010477.1 - 1 474 3154433

View Full Product Details