Gene Transcripts

Gene Symbol
SUB1
Entrez Gene ID
10923
Gene Name
SUB1 homolog, transcriptional regulator
Gene Aliases
P15, PC4, p14
Chromosome Location
Chr. 5: 32531778 - 32604079 on Build GRCh38
UniGene ID
Hs.229641
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006713.3 NP_006704.3 3 216
XM_011513944.2 XP_011512246.1 4 469
XM_017008986.1 XP_016864475.1 4 312
XM_017008987.1 XP_016864476.1 4 189
GenBank mRNA AB209285.1 - 3 111
AK294124.1 - 3 158
AK307268.1 - 3 148
AK311754.1 - 3 158
BC009610.1 - 3 144 4297490
BC010537.1 - 3 119 3457167
BC018189.1 - 3 138 3881373
BC022339.1 - 3 149 4134117
BC063402.1 - 3 132
BC071655.1 - 3 133
BC105972.1 - 3 114 3907729
BX472106.1 - 3 217
CR456902.1 - 2 88
U12979.1 - 2 88
X79805.1 - 2 88

View Full Product Details