Gene Transcripts

Gene Symbol
Cyb5b
Entrez Gene ID
66427
Gene Name
cytochrome b5 type B
Gene Aliases
1810044O22Rik, AU015618, Cyb5m
Chromosome Location
Chr. 8: 107150661 - 107187470 on Build GRCm38
UniGene ID
Mm.20242
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025558.5 NP_079834.2 2 323
GenBank mRNA AK003333.2 - 2 324
AK003815.1 - 2 313
AK007780.1 - 343
AK019263.1 - 2 323
AK052810.1 - 2 324
AK083359.1 - 2 169
AK088970.1 - 2 325
AK165543.1 - 2 327
AK166544.1 - 2 301
AK168422.1 - 2 313
BC046479.1 - 2 286
BC046810.1 - 2 289
BC048147.1 - 2 313
BC054749.1 - 2 344 6410090
BC058812.1 - 2 286 6530835
BC062980.1 - 2 335 6842939

View Full Product Details