Gene Transcripts

Gene Symbol
Lix1
Entrez Gene ID
66643
Gene Name
limb expression 1 homolog (chicken)
Gene Aliases
5730466L18Rik
Chromosome Location
Chr. 17: 17402686 - 17459387 on Build GRCm38
UniGene ID
Mm.268018
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025681.2 NP_079957.2 2 494
GenBank mRNA AF351204.1 - 2 235
AK012220.1 - 2 302
AK017685.1 - 2 313
BC049574.1 - 2 508 6593108
BC052700.1 - 2 294 6416835
BC063057.1 - 2 287 5715784

View Full Product Details