Gene Transcripts

Gene Symbol
DRD5
Entrez Gene ID
1816
Gene Name
dopamine receptor D5
Gene Aliases
DBDR, DRD1B, DRD1L2
Chromosome Location
Chr. 4: 9781634 - 9784009 on Build GRCh38
UniGene ID
Hs.380681
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000798.4 NP_000789.1 1 1283
GenBank mRNA AK313897.1 - 1 1110
AY136750.1 - 1 887
BC009748.2 - 1 938 3928370

View Full Product Details