Gene Transcripts

Gene Symbol
Wtap
Entrez Gene ID
60532
Gene Name
Wilms tumour 1-associating protein
Gene Aliases
2810408K05Rik, 9430038B09Rik
Chromosome Location
Chr. 17: 12966798 - 12992808 on Build GRCm38
UniGene ID
Mm.275521
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113532.1 NP_001107004.1 6 721
NM_175394.2 NP_780603.1 6 769
GenBank mRNA AK031965.1 - 6 778
AK076111.1 - 6 763
AK220249.1 - 2 376
BC046416.1 - 6 650
BC047338.1 - 6 650
BC093504.1 - 6 654

View Full Product Details