Gene Transcripts

Gene Symbol
Rgs6
Entrez Gene ID
50779
Gene Name
regulator of G-protein signaling 6
Gene Aliases
Chromosome Location
Chr. 12: 82616176 - 83158815 on Build GRCm38
UniGene ID
Mm.153013
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282061.1 NP_001268990.1 11 841
NM_015812.3 NP_056627.1 11 1123
GenBank mRNA AF061933.1 - 8 568
AK052527.1 - 11 1124
AK053087.1 - 11 842
AK139323.1 - 11 1238
BC038616.1 - 9 668
BC125029.1 - 11 827 40062355
BC125030.1 - 11 827 40062356

View Full Product Details