Gene Transcripts

Gene Symbol
Samhd1
Entrez Gene ID
56045
Gene Name
SAM domain and HD domain, 1
Gene Aliases
E330031J07Rik, Mg11
Chromosome Location
Chr. 2: 157097529 - 157135277 on Build GRCm38
UniGene ID
Mm.248478
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001139520.1 NP_001132992.1 10, 11 1317
NM_018851.3 NP_061339.3 10, 11 1317
GenBank mRNA AK054490.1 - 10, 11 1216
AK078936.1 - 10, 11 1260
AK128954.1 - 9, 10 1115
AK151335.1 - 10, 11 1191
AK153390.1 - 10, 11 1191
AK170688.1 - 10, 11 1216
AK171565.1 - 10, 11 1217
BC012721.1 - 10, 11 1254 4037046
BC067198.1 - 10, 11 1240 6826682
U15635.1 - 10, 11 1241

View Full Product Details