Gene Transcripts

Gene Symbol
Ddx21
Entrez Gene ID
56200
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 21
Gene Aliases
AI255159, AL022742, D10Ertd645e, D10Wsu42e
Chromosome Location
Chr. 10: 62580247 - 62602298 on Build GRCm38
UniGene ID
Mm.413275
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019553.2 NP_062426.2 6 1289
GenBank mRNA AF159131.3 - 6 1250
AK010281.1 - 6 1169
AK145578.1 - 6 1273
AK160095.1 - 6 1263
BC030895.1 - 5 721
BC043655.1 - 6 1261 5363948
BC059237.1 - 6 1249 6412782
BC060220.1 - 6 1241

View Full Product Details