Gene Transcripts
- Gene Symbol
- EBNA1BP2
- Entrez Gene ID
- 10969
- Gene Name
- EBNA1 binding protein 2
- Gene Aliases
- EBP2, NOBP, P40
- Chromosome Location
- Chr. 1: 43164174 - 43172570 on Build GRCh38
- UniGene ID
- Hs.346868
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001159936.1 |
NP_001153408.1
|
3, 4 |
457 |
|
NM_006824.2 |
NP_006815.2
|
2, 3 |
340 |
|
GenBank mRNA |
AB451256.1 |
- |
2, 3 |
143 |
|
AB451381.1 |
- |
2, 3 |
143 |
|
AK223455.1 |
- |
2, 3 |
323 |
|
AK295006.1 |
- |
2, 3 |
327 |
|
AK299476.1 |
- |
2, 3 |
325 |
|
AK307440.1 |
- |
2, 3 |
286 |
|
BC009175.2 |
- |
2, 3 |
281 |
2966681 |
BC073924.1 |
- |
8, 9 |
692 |
|
BM791254.1 |
- |
2, 3 |
341 |
|
BT006920.1 |
- |
2, 3 |
143 |
|
CN390915.1 |
- |
3, 4 |
475 |
|
CR456975.1 |
- |
2, 3 |
143 |
|
U86602.1 |
- |
2, 3 |
285 |
|
View Full Product Details