Gene Transcripts

Gene Symbol
Rnf32
Entrez Gene ID
56874
Gene Name
ring finger protein 32
Gene Aliases
1700009J01Rik, 2700025B22Rik, 4930542N22Rik, Lmbr2
Chromosome Location
Chr. 5: 29195170 - 29225526 on Build GRCm38
UniGene ID
Mm.487302
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289757.1 NP_001276686.1 4 343
NM_021470.6 NP_067445.3 4 315
GenBank mRNA AF190664.1 - 1 77
AK005795.1 - 4 270
AK006379.1 - 4 259
AK012288.1 - 4 343
AK016026.1 - 4 286
BC050964.1 - 4 258
BC061103.1 - 4 281
BC089493.1 - 4 258 6741898
BY293373.1 - 3, 4 263

View Full Product Details