Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144523.2 | NP_653106.1 | 2 | 975 | |
GenBank mRNA | AK031232.1 | - | 2 | 975 | |
AK149106.1 | - | 2 | 906 | ||
AK154951.1 | - | 2 | 901 | ||
AK170489.1 | - | 2 | 974 | ||
BC006964.1 | - | 2 | 880 | 3153893 |