Gene Transcripts

Gene Symbol
Mtg2
Entrez Gene ID
52856
Gene Name
mitochondrial ribosome associated GTPase 2
Gene Aliases
1810011P19Rik, 2900056P18Rik, D2Bwg0647e, Gtpbp5
Chromosome Location
Chr. 2: 180070593 - 180085902 on Build GRCm38
UniGene ID
Mm.219658
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083328.1 NP_001076797.1 5 428
NM_001083329.1 NP_001076798.1 4 334
NM_028422.1 NP_082698.1 5 473
NM_181424.3 NP_852089.2 6 567
GenBank mRNA AK040689.1 - 5 430
AK201295.1 - 2 139
AW647214.1 - 3 298
BC025144.1 - 6 566 4483063
BC028518.1 - 3 312
DT929445.1 - 2 51

View Full Product Details