Gene Transcripts

Gene Symbol
DIAPH2
Entrez Gene ID
1730
Gene Name
diaphanous related formin 2
Gene Aliases
DIA, DIA2, DRF2, POF, POF2
Chromosome Location
Chr. X: 96684663 - 97600598 on Build GRCh38
UniGene ID
Hs.226483
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006729.4 NP_006720.1 9 1350
NM_007309.3 NP_009293.1 9 1350
GenBank mRNA AK291272.1 - 9 1232
BC117414.1 - 10 1017 40125965
BC143838.1 - 10 1017 9052353
Y15908.1 - 9 1304
Y15909.1 - 9 1304

View Full Product Details