Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009315.3 | NP_033341.1 | 4, 5 | 482 | |
GenBank mRNA | AK153731.1 | - | 530 | ||
AK154867.1 | - | 4, 5 | 515 | ||
AK213626.1 | - | 1, 2 | 102 | ||
AK216752.1 | - | 1, 2 | 102 | ||
BC058583.1 | - | 4, 5 | 482 | 6514960 | |
D49439.1 | - | 3, 4 | 415 |