Gene Transcripts

Gene Symbol
Tgm3
Entrez Gene ID
21818
Gene Name
transglutaminase 3, E polypeptide
Gene Aliases
AI893889, TG E, TG(E), TGE, TGase E, TGase-3
Chromosome Location
Chr. 2: 129987794 - 130050399 on Build GRCm38
UniGene ID
Mm.2961
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009374.3 NP_033400.2 4 571
GenBank mRNA BC129890.1 - 4 537
BC129891.1 - 4 537 40111493
L10385.1 - 4 562

View Full Product Details