Gene Transcripts

Gene Symbol
Tnfsf10
Entrez Gene ID
22035
Gene Name
tumor necrosis factor (ligand) superfamily, member 10
Gene Aliases
A330042I21Rik, AI448571, APO-2L, Ly81, TL2, Trail
Chromosome Location
Chr. 3: 27317077 - 27339665 on Build GRCm38
UniGene ID
Mm.1062
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009425.2 NP_033451.1 5 752
GenBank mRNA AK142960.1 - 5 754
AK153581.1 - 5 802
AK157633.1 - 753
BC011469.1 - 5 718
BC138807.1 - 5 726 8861829
BC138808.1 - 5 726 8861830
U37522.1 - 5 728

View Full Product Details