Gene Transcripts

Gene Symbol
Usf1
Entrez Gene ID
22278
Gene Name
upstream transcription factor 1
Gene Aliases
bHLHb11
Chromosome Location
Chr. 1: 171411304 - 171418759 on Build GRCm38
UniGene ID
Mm.8
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF479773.1 - 2 66
AK142437.1 - 4 288
BC049784.1 - 4 231 6506829
CJ050979.1 - 4 367
DV653810.1 - 4 193

View Full Product Details