Gene Transcripts

Gene Symbol
Timm17b
Entrez Gene ID
21855
Gene Name
translocase of inner mitochondrial membrane 17b
Gene Aliases
17kDa, Sfc3, mTim17b
Chromosome Location
Chr. X: 7899242 - 7907652 on Build GRCm38
UniGene ID
Mm.27393
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011591.4 NP_035721.1 6 1286
GenBank mRNA AK049584.1 - 6 1287

View Full Product Details