Gene Transcripts
- Gene Symbol
- Zfp148
- Entrez Gene ID
- 22661
- Gene Name
- zinc finger protein 148
- Gene Aliases
- 2210405J08Rik, AI480666, AW045217, BERF-1, BFCOL1, ZBP-89, Znf148
- Chromosome Location
- Chr. 16: 33380775 - 33503903 on Build GRCm38
- UniGene ID
- Mm.392667
- Species
- Mus musculus
- Gene Symbol
- C330025O08Rik
- Entrez Gene ID
- 78698
- Gene Name
- RIKEN cDNA C330025O08 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.348587
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK021224.1 |
- |
1 |
200 |
|
View Full Product Details